Skip to Content
Merck
All Photos(4)

Key Documents

B9300

Sigma-Aldrich

Benzophenone

ReagentPlus®, 99%

Synonym(s):

Diphenyl ketone, Diphenylmethanone, NSC 8077

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2CO
CAS Number:
Molecular Weight:
182.22
Beilstein:
1238185
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

1 mmHg ( 108 °C)

Quality Level

product line

ReagentPlus®

Assay

99%

form

flakes

bp

305 °C (lit.)

mp

47-51 °C (lit.)

functional group

ketone
phenyl

SMILES string

O=C(C1=CC=CC=C1)C2=CC=CC=C2

InChI

1S/C13H10O/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H

InChI key

RWCCWEUUXYIKHB-UHFFFAOYSA-N

Gene Information

rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Photo-crosslinkable bioglue development: A study demonstrated the creation of a surface-independent bioglue using a photo-crosslinkable benzophenone moiety, potentially enhancing medical adhesives for diverse applications (Shi et al., 2024).

  • Organic explosive trace analysis: Research highlighted the use of benzophenone for improved preconcentration and analysis of organic explosive traces in aqueous samples, advancing forensic methodologies (Setayeshfar et al., 2024).

  • OLEDs blue emitters: A publication introduced bicarbazole-benzophenone based twisted donor-acceptor derivatives as potential blue TADF emitters for OLED applications, showcasing the versatility of benzophenone in electronic devices (Siddiqui et al., 2024).

  • Photodynamic antibacterial nonwovens: Research focused on the modification of traditional composite nonwovens with benzophenone to enable stable storage of light absorption transients and enhance photodynamic antibacterial effects, contributing to medical textile innovations (Li et al., 2024).

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - STOT RE 2 Oral

Target Organs

Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

280.4 °F - closed cup

Flash Point(C)

138 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jiawei Yang et al.
ACS applied materials & interfaces, 11(27), 24802-24811 (2019-06-14)
Recent innovations highlight the integration of diverse materials with synthetic and biological hydrogels. Examples include brain-machine interfaces, tissue regeneration, and soft ionic devices. Existing methods of strong adhesion mostly focus on the chemistry of bonds and the mechanics of dissipation
Claire Chatalova-Sazepin et al.
Angewandte Chemie (International ed. in English), 54(18), 5443-5446 (2015-03-05)
A three-component carboetherification of unactivated alkenes has been developed allowing the rapid building of complexity from simple starting materials. A wide range of α-substituted styrenes underwent smooth reactions with unactivated alkyl nitriles and alcohols to afford γ-alkoxy alkyl nitriles with
Masayuki Kyomoto et al.
Biomaterials, 34(32), 7829-7839 (2013-07-31)
We investigated the production of free radicals on a poly(ether-ether-ketone) (PEEK) substrate under ultraviolet (UV) irradiation. The amount of the ketyl radicals produced from the benzophenone (BP) units in the PEEK molecular structure initially increased rapidly and then became almost
L L Barnkob et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 30(2), 395-402 (2012-11-17)
The aim of this study was to investigate the effect of relative humidity on the migration of benzophenone from paperboard into the food simulant Tenax®. Kinetic migration investigations were carried out with three relative humidities in the interval between 39%
Feifan Yu et al.
PloS one, 8(2), e56597-e56597 (2013-02-21)
Affinity proteins binding to antibody constant regions have proved to be invaluable tools in biotechnology. Here, protein engineering was used to expand the repertoire of available immunoglobulin binding proteins via improvement of the binding strength between the widely used staphylococcal

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service