Skip to Content
Merck
All Photos(2)

Key Documents

132551

Sigma-Aldrich

1,2-Diaminocyclohexane, mixture of cis and trans

99%

Synonym(s):

1,2-Cyclohexanediamine, DHC 99

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H10(NH2)2
CAS Number:
Molecular Weight:
114.19
Beilstein:
506142
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.4 mmHg ( 20 °C)

Assay

99%

form

liquid

refractive index

n20/D 1.49 (lit.)

bp

92-93 °C/18 mmHg (lit.)

density

0.931 g/mL at 25 °C (lit.)

SMILES string

NC1CCCCC1N

InChI

1S/C6H14N2/c7-5-3-1-2-4-6(5)8/h5-6H,1-4,7-8H2

InChI key

SSJXIUAHEKJCMH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

1,2-Diaminocyclohexane undergoes non-templated reaction with homochiral as well as the racemic form of trans-1,2-diaminocyclohexane with terephthaldehyde to yield (3+3)-cyclocondensed molecular triangles. It acts as ligand and forms organotin complexes, having potential applications as metal-based antitumour drugs.

Application

1,2-Diaminocyclohexane was used in the synthesis of chiral ruthenium(IV)-oxo complexes.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J J Bonire et al.
Journal of inorganic biochemistry, 83(2-3), 217-221 (2001-03-10)
Platinum compounds containing the ligand 1,2-diaminocyclohexane (DACH) such as tetraplatin [PtCl4(DACH)] have been found to be active in cisplatin-resistant tumour models. In an attempt to develop novel metal-based drugs with a different therapeutic profile to cisplatin, we have synthesised a
(3+ 3)-Cyclocondensation of the enantiopure and racemic forms of trans-1, 2-diaminocyclohexane with terephthaldehyde. Formation of diastereomeric molecular triangles and their stereoselective solid-state stacking into microporous chiral columns.
Chadim M, et al.
Tetrahedron Asymmetry, 12(1), 127-133 (2001)
Chiral ruthenium (IV)-oxo complexes. Structure, reactivities of [Ru (terpy)(Nn N) O]2+Nn N= N, N, N', N'-tetramethyl-1, 2-diaminocyclohexane) and [Ru (Me3 tacn)(cbpy) O]2+ (cbpy=(-)-3, 3'-[(4 S-trans)-1, 3-dioxolane-4, 5-dimethyl]-2, 2'-bipyridine).
Cheng WC, et al.
Inorgorganica Chimica Acta, 242(1), 105-113 (1996)
Yu He et al.
Inorganic chemistry, 50(24), 12651-12660 (2011-11-11)
Iron complexes with the tetradentate N-donor ligand N,N'-di(phenylmethyl)-N,N'-bis(2-pyridinylmethyl)-1,2-cyclohexanediamine (bbpc) are reported. Despite the benzyl groups present on the amines, the iron compounds catalyze the oxygenation of cyclohexane to an extent similar to those employing less sterically encumbered ligands. The catalytic
Robert Häner et al.
Journal of the American Chemical Society, 132(21), 7466-7471 (2010-05-13)
Oligopyrenotides, abiotic oligomers that exhibit significant structural analogies to the nucleic acids, are described. They are composed of achiral, phosphodiester-linked pyrene building blocks and a single chiral 1,2-diaminocyclohexane unit. These oligomers form stable hybrids in aqueous solution. Hybridization is based

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service