Skip to Content
Merck
All Photos(1)

Documents

D170208

Sigma-Aldrich

1,3-Dimethylnaphthalene

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H6(CH3)2
CAS Number:
Molecular Weight:
156.22
Beilstein:
2039378
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

liquid

refractive index

n20/D 1.609 (lit.)

bp

263 °C (lit.)

density

0.982 g/mL at 25 °C (lit.)

SMILES string

Cc1cc(C)c2ccccc2c1

InChI

1S/C12H12/c1-9-7-10(2)12-6-4-3-5-11(12)8-9/h3-8H,1-2H3

InChI key

QHJMFSMPSZREIF-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Odd G Brakstad et al.
Chemosphere, 204, 87-91 (2018-04-14)
Hydrocarbon biodegradation may be slower in cold Arctic than in temperate seawater, and this will affect the toxicity time window of the hydrocarbons. In this study, the acute toxicities of water-soluble phases of 1,3-dimethylnaphthalene, phenanthrene, fluoranthene, and low energy water-accommodated
Tjalling Jager et al.
Environmental science & technology, 51(13), 7707-7713 (2017-06-10)
Efficiently assessing and managing the risks of pollution in the marine environment requires mechanistic models for toxic effects. The General Unified Threshold model for Survival (GUTS) provides a framework for deriving toxicokinetic-toxicodynamic (TKTD) models for the end point survival. Two
Xu-Hui Huang et al.
Food & function, 12(4), 1626-1638 (2021-01-22)
Clam is a kind of nutritious, delicious and economical aquatic food around the world and is famous for its unique aroma. Instrumental analysis, sensory analysis, and comprehensive statistical analysis were performed to explain the relationship between aroma and odorants in
Sangwoo Lee et al.
Environmental science. Processes & impacts, 19(9), 1117-1125 (2017-08-08)
Polycyclic aromatic hydrocarbons (PAHs) and alkylated PAHs are known to be major toxic contaminants in spills of petroleum hydrocarbons (oil). Spilled oil undergoes weathering and over time, PAHs go through a series of compositional changes. PAHs can disrupt endocrine functions
Kunal Roy et al.
European journal of medicinal chemistry, 44(5), 1941-1951 (2008-12-27)
A series of naphthalene and non-naphthalene derivatives (n=42) having cytochrome P450 2A6 and 2A5 inhibitory activities, reported by Rahnasto et al., were subjected to QSAR and QAAR studies. The analyses were performed using electronic, spatial, shape and thermodynamic descriptors to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service