Skip to Content
Merck
All Photos(1)

Documents

13618

Sigma-Aldrich

Wang resin

extent of labeling: 0.6-1.2 mmol/g loading

Synonym(s):

p-Alkoxy-benzyl alcohol polymer-bound, p-Alkoxybenzyl alcohol resin, [4-(Hydroxymethyl)phenoxymethyl]polystyrene

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

extent of labeling

0.6-1.2 mmol/g loading

matrix

crosslinked with 1% DVB

particle size

200-400 mesh

functional group

alcohol

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service