All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C5H8N2
CAS Number:
Molecular Weight:
96.13
Beilstein:
106381
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
form
solid
bp
268 °C (lit.)
mp
78-81 °C (lit.)
SMILES string
CCc1ncc[nH]1
InChI
1S/C5H8N2/c1-2-5-6-3-4-7-5/h3-4H,2H2,1H3,(H,6,7)
InChI key
PQAMFDRRWURCFQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Microwave plasma reaction of 2-ethylimidazole on poly(dimethylsiloxane) surface has been investigated.
2-Ethylimidazole is used as a ligand for the synthesis of hydrophobic cobalt-ethylimidazolate frameworks.
2-Ethylimidazole is used as a ligand for the synthesis of hydrophobic cobalt-ethylimidazolate frameworks.
Application
2-Ethylimidazole has been used as internal standard in the quantification of 4-(5-)methylimidazole (4MI) in coffee by GC-MS method.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Jangho Lee et al.
Journal of separation science, 41(17), 3415-3423 (2018-07-20)
A rapid analytical method was developed for the determination of 4-methylimidazole from red ginseng products containing caramel colors by using dispersive liquid-liquid microextraction with in situ derivatization followed by gas chromatography with mass spectrometry. Chloroform and acetonitrile were selected as
Min-Chul Jung et al.
Journal of food science, 82(10), 2526-2531 (2017-09-12)
The objective of this study was to determine the reduction of 4(5)-methylimidazole (4-MI) under various baking conditions. For 4-MI analysis, an analytical method using gas chromatography-mass spectrometry was developed. The developed method was validated with linearity (r A consumer awareness
Hydrophobic cobalt-ethylimidazolate frameworks: Phase-pure syntheses and possible application in cleaning of contaminated water
Bhadra BN, et al.
Inorganic Chemistry, 55, 11362-11371 (2016)
Sumin Lee et al.
Journal of food science, 81(1), T262-T267 (2015-12-15)
In this study, the effect of food additives such as iron sulfate, magnesium sulfate, zinc sulfate, citric acid, gallic acid, and ascorbic acid on the reduction of 4(5)-methylimidazole (4(5)-MI) was investigated using a soy sauce model system. The concentration of
Wen-Huan Huang et al.
Nanoscale, 12(38), 19804-19813 (2020-09-24)
The hydrogen evolution reaction is a key half reaction for water electrolysis and is of great significance. Pt-based nanomaterials are promising candidates for HER electrocatalysts. However, the high price of platinum and poor durability impede their practical applications. Herein, a
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service