Skip to Content
Merck
All Photos(1)

Key Documents

M79204

Sigma-Aldrich

N-Methylpyrrolidine

97%

Synonym(s):

1-Methylpyrrolidine, N-Methylpyrrolidine, N-Methyltetrahydropyrrole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11N
CAS Number:
Molecular Weight:
85.15
Beilstein:
102445
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

bp

80-81 °C (lit.)

density

0.819 g/mL at 25 °C (lit.)

SMILES string

CN1CCCC1

InChI

1S/C5H11N/c1-6-4-2-3-5-6/h2-5H2,1H3

InChI key

AVFZOVWCLRSYKC-UHFFFAOYSA-N

Application

N-Methylpyrrolidine can be used as a starting material to synthesize:
N-methyl pyrrolidine-zinc borohydride (ZBHNMP), a reducing agent for reduction of aldehydes, ketones, acid chlorides, and esters. ZBHNMP is also used in the reductive amination of aldehydes and ketones to their corresponding amines.
N-alkyl-N-methyl-pyrrolidinium bis(trifluoromethanesulfonyl)imide ionic liquids.
N-methylpyrrolidine-2-one hydrotribromide applicable as a catalyst for aziridination of alkenes.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Flam. Liq. 2 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-0.4 °F - closed cup

Flash Point(C)

-18 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of the alkyl group on the synthesis and the electrochemical properties of N-alkyl-N-methyl-pyrrolidinium bis (trifluoromethanesulfonyl) imide ionic liquids
Appetecchi GB, et al.
Electrochimica Acta, 54(4), 1325-1332 (2009)
Reductive amination of aldehydes and ketones to their corresponding amines with N-methylpyrrolidine zinc borohydride
Alinezhad H, et al.
Tetrahedron Letters, 50(6), 659-661 (2009)
N-methylpyrrolidine-2-one hydrotribromide: An efficient and new catalyst for the aziridination of alkenes using Chloramine-T under solvent free conditions
Jain SL, et al.
J. Mol. Catal. A: Chem., 256(1-2), 16-20 (2006)
N-Methylpyrrolidine-zinc borohydride: As a new stable and efficient reducing agent in organic synthesis
Tajbakhsh M, et al.
Synthetic Communications, 33(2), 229-236 (2003)
Kenjiro Ono et al.
Biological psychiatry, 52(9), 880-886 (2002-10-26)
Cerebral deposition of amyloid beta-peptide (Abeta) is a major neuropathologic feature in Alzheimer's disease (AD). A consistent protective effect of smoking on AD has been documented by many case-control studies. It has been suggested that nicotine, a major component of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service