Skip to Content
Merck
All Photos(2)

Key Documents

116238

Sigma-Aldrich

5,7-Dimethoxycoumarin

98%

Synonym(s):

Citropten, Limettin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H10O4
CAS Number:
Molecular Weight:
206.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

mp

146-149 °C (lit.)

SMILES string

COc1cc(OC)c2C=CC(=O)Oc2c1

InChI

1S/C11H10O4/c1-13-7-5-9(14-2)8-3-4-11(12)15-10(8)6-7/h3-6H,1-2H3

InChI key

NXJCRELRQHZBQA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5,7-dimethoxycoumarin is isolated and identified from leaves and fruits of Pelea anisata H. Mann, a plant whose fruit are used in the construction of mohikana leis. It induces frameshift mutagenesis in bacteria. It also causes lethal photosensitization and the formation of sister chromatid exchanges in Chinese hamster cells.

Biochem/physiol Actions

5,7-Dimethoxycoumarin induces the processes of differentiation and melanogenesis in murine (B16) and human (A375).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniela Alesiani et al.
International journal of oncology, 34(6), 1727-1735 (2009-05-09)
In this study, the processes of differentiation and melanogenesis induced by 5,7-dimethoxycoumarin in murine (B16) and human (A375) melanoma cells were investigated. Taking into account the previously demonstrated antiproliferative and differentiation activities of this compound, we examined Ras/Raf/Mek/Erk mitogen-activated protein
Takeshi Kinoshita et al.
Chemical & pharmaceutical bulletin, 50(1), 118-120 (2002-02-05)
A new C-8 prenylated 5,7-dimethoxycoumarin named omphamurrayin was isolated from the leaves of Murraya paniculata var. omphalocarpa, and its structure was established as 5,7-dimethoxy-8-(1-oxo-2-senecioyl-3-methyl-3-butenyl)-2H-1-benzopyran-2-one on the basis of the spectroscopic evidence. The taxonomic status of M. paniculata var. omphalocarpa is
Isolation and characterization of the photoadducts of 5,7-dimethoxycoumarin and adenosine.
T H Cho et al.
Photochemistry and photobiology, 46(2), 305-309 (1987-08-01)
V Jung et al.
Biochimica et biophysica acta, 740(1), 64-72 (1983-05-20)
The photobinding of 5,7-dimethoxycoumarin to isolated adenovirus-type 2 DNA has been investigated with respect to the influence of the ionic environment, and varying molar ratios of DNA(p): 5,7-dimethoxycoumarin. In particular, the ultraviolet radiation-induced covalent addition of 5,7-dimethoxycoumarin to adenovirus DNA
P Treffel et al.
Skin pharmacology : the official journal of the Skin Pharmacology Society, 5(2), 108-113 (1992-01-01)
It is a general rule that percutaneous absorption is increased when the site of application is occluded. In this study we compared the in vitro permeation profiles of two molecules with different physicochemical properties under occluded versus unoccluded conditions. Human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service