Skip to Content
Merck
All Photos(1)

Key Documents

178373

Sigma-Aldrich

2′,4′-Dichloroacetophenone

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3COCH3
CAS Number:
Molecular Weight:
189.04
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

refractive index

n20/D 1.5635 (lit.)

bp

140-150 °C/15 mmHg (lit.)

mp

33-34 °C (lit.)

SMILES string

CC(=O)c1ccc(Cl)cc1Cl

InChI

1S/C8H6Cl2O/c1-5(11)7-3-2-6(9)4-8(7)10/h2-4H,1H3

InChI key

XMCRWEBERCXJCH-UHFFFAOYSA-N

Related Categories

Application

2′,4′-Dichloroacetophenone was used in the synthesis of the Schiff base derivatives.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microwave assisted rapid and efficient synthesis of nitrogen and sulphur containing heterocyclic compounds and their pharmacological evaluation.
Indian J. Chem. B, 45(7), 1762-1762 (2006)
Xue-yan Shi et al.
Se pu = Chinese journal of chromatography, 20(1), 34-36 (2003-01-25)
A new cyclodextrin (CD) derivative, 2,6-di-O-benzyl-3-O-valeryl-beta-CD, was synthesized and characterized by 1H NMR and IR. Using the beta-CD derivative as chiral stationary phase of capillary gas chromatography, one chiral column was prepared. On this column, the enantiomeric excesses (e.e.) of
A Brundin et al.
Biochemical pharmacology, 31(23), 3885-3890 (1982-12-01)
alpha-Haloketones are highly reactive compounds, which are known to undergo enzymatic reduction to methyl ketones. The objective of this research was to characterize the enzymes involved in this reaction and to investigate the mechanism of the reaction. 2,2',4'-Trichloroacetophenone was reduced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service