Skip to Content
Merck
All Photos(1)

Key Documents

111589

Sigma-Aldrich

1,6-Heptadien-4-ol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCH2CH(OH)CH2CH=CH2
CAS Number:
Molecular Weight:
112.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.45 (lit.)

bp

151 °C (lit.)

density

0.864 g/mL at 25 °C (lit.)

functional group

allyl
hydroxyl

SMILES string

OC(CC=C)CC=C

InChI

1S/C7H12O/c1-3-5-7(8)6-4-2/h3-4,7-8H,1-2,5-6H2

InChI key

UTGFOWQYZKTZTN-UHFFFAOYSA-N

General description

1,6-Heptadien-4-ol can help in synthesizing the derivatives of guanine, adenine, uracil and thymine via Mitsunobu condensation.

Application

1,6-Heptadien-4-ol (monoterpene alcohol) was used as the internal standard in the quantitation and identification of the important aroma components in wine. It was used to study the co- and terpolymerization reactions of carbon monoxide and propene with dicationic biphosphine palladium(II) as the catalyst.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jui-Yi Tsai et al.
The Journal of organic chemistry, 68(4), 1235-1241 (2003-02-15)
A series of 1,6-heptadienes, substituted in the 4 position with nucleic acid bases 1-6, have been synthesized via Mitsunobu condensations. Guanine, adenine, thymine, and uracil derivatives can be prepared directly by coupling the protected base with 1,6-heptadien-4-ol (7). However, coupling
Magdalena Skotniczny et al.
Foods (Basel, Switzerland), 9(8) (2020-08-08)
The influence of fruit varieties on yeast ecology during spontaneous plum mash fermentation was investigated. Yeast colonies were isolated from mashes obtained from four plum varieties throughout fermentation in laboratory conditions during two consecutive years. The yeast strains were differentiated
Validation of a Solid-Phase Microextraction Method for Headspace Analysis of Wine Aroma Components.
American Journal of Enology and Viticulture, 56(1), 37-45 (2005)
Tivadar Feczkó et al.
International journal of pharmaceutics, 562, 333-341 (2019-03-15)
Vinyl alcohol (VA) copolymers having fine tunable polarities are emerging materials in drug delivery applications. VA copolymers rendering well-defined molecular architecture (C/OH ratio = 2, 4, 5 and 8) were used as carriers for model drug compound, fluorescein, which exhibited significantly different
Copolymerization of carbon monoxide with exo-methylenecycloalkane and dienes: synthesis of functionalized aliphatic polyketones.
Kettunen M, et al.
Polymer International, 50(11), 1223-1227 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service