MilliporeSigma
All Photos(4)
Key Documents
MISC-MP4K-POLY-WUV-IRDUAL-C4
Zarbeco, LLC
Zarbeco MiScope Megapixel MP4K
with Polyspectral Imaging and white, ultraviolet (UV), and two infrared (IR) LEDs
Synonym(s):
portable digital microscope, Handheld digital microcope, USB digital microscope, Fluorescence microscope
About This Item
PDF Icon
Shipped & Sold By Zarbeco, LLC
A Trusted Marketplace Seller
PDF Icon
Have a question?
manufacturer/tradename
Zarbeco, LLC
General description
MiScope® Megapixel Polyspectral MP4K WUVIRdual (MISC-MP4K-POLY-WUV-IRdual) is a superior grade, lab quality, portable digital microscope.  It includes white, ultraviolet (UV) and two wavelength infrared (IR) LEDs for polyspectral imaging with blended lighting.  With its' micron level resolution, small footprint, easy to use included software, portability, and remote interface, it is a useful tool to supplement or even replace conventional stereo microscopes. Use your MIScope on-the-go or on valuable lab bench space and bring it to your sample, place it on top of it, and still pictures and movies can easily be saved, annotated, measured, drawn-on and enhanced with the included Video ToolBox Premier PC software. Powered by the USB-3 port of your laptop, desktop, or mobile device* it can go anywhere you need high quality images. Assembled in New Jersey by Zarbeco, LLC established in 2001.
  • 13 Megapixel camera
  • Resolution to 1.1 microns
  • 11x-360x magnification
  • Connects to the USB 3.0 port of your desktop or laptop computer, or mobile device*
  • Includes our Video Toolbox Premier PC Software for capturing still and moving images, for all PRO features plus side by side comparison, calibrated overlays, low-light frame averaging, enhanced measurement, pixel-level adjustment, magnifier window, and edge detection (MAC software optional MiTools on Mac App store).
  • Comes with variable Ushims for intermediate magnification.
  • Can be about 6-8 inches away from your object to see 1.9 inch field of view.
  • Move the arm on the side of your MiScope Megapixel MP4K and it will focus and zoom at the same time.
  • When connected to the USB 3.0 port of your computer, it has fast image speeds at up to 60 frames per second without lag; 30 frames per second at 4K resolution with barely perceptible lag.
  • Has white LEDs with on-off switch to use side or bottom lighting with optional accessories. 
  • Has Ultraviolet (UV) and Infrared (IR) LEDs for fluorescent microscopy and true polyspectral imaging with blended lighting and combined wavelengths. 
*Apple iPad and iPhone need optional adapter. Apple, Mac, iPhone, and iPad are trademarks of Apple Inc., registered in the U.S. and other countries.  
Warning: Infrared and Ultraviolet light is harmful.  Do not expose eyes or skin to rays. Wear UV protective eyewear and protective clothing to prevent exposure.  
These units are not recommended for unsupervised use by children.
Legal Information
MiScope is a registered trademark of Zarbeco, LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)