Zarbeco, LLC
Zarbeco has manufactured digital optical microscopes, cameras, and imaging software in the USA since 2001. Our popular MiScope handheld digital microscopes, with the options for ultraviolet (UV) and infrared (IR) illumination, allows you to capture still and moving images and do precision measurements on your desktop, laptop or mobile device. Our professional products are used throughout the world at Fortune 500 companies, government agencies, and universities.
Personalized Customer Service
We offer personalized customer service in-house in New Jersey from 10 AM to 6 PM EST.
In-house Support
Our Video ToolBox line of software, which comes with our portable digital microscopes, offers outstanding customer support.
Contact Zarbeco, LLC
Phone:
973-933-2043
Email:
Fax:
973-933-2336
Address:
52 Main Street
STE 1
Succasunna, NJ, 07876
USA
STE 1
Succasunna, NJ, 07876
USA
MiScope Portable Digital Microscopes
Zarbeco has sold over 12,000 MiScope handheld digital microscopes worldwide for use in industries such as scientific labs, forensic science, entomology, art conservation, and manufacturing. The MiScope portable digital microscope provides a quick, compact way to inspect samples while taking up a small amount of space on a lab bench or desk. The live images can be used on a computer, tablet or phone. The handheld microscopes include built-in white lights, with the option to add ultraviolet and infrared LEDs.
MiScope MP3 & MiScope MP4K
The MiScope Megapixel MP3 is a scientific quality portable digital microscope with 3.4 megapixels, magnification up to 180x and less than 1.7 micron resolution. It is USB3 powered and images at video frame rates up to 60 frames per second. The MiScope-MP3 includes Video Toolbox PRO PC software with image and video capture options (including time lapse recording) as well as tools for live video annotation and measurement. The handheld digital microscope can be used to view samples such as slides, fibers, and insects.
The MiScope Megapixel MP4K is a premium quality portable digital microscope with 13 megapixels, magnification up to 360x and resolution to 1.1 microns. The included Video Toolbox Premier software adds advanced viewing and capture features, additional measurement tools, edge detection and calibrated overlays.