MilliporeSigma
All Photos(4)
Key Documents
MISC-MP4K-C4
Zarbeco, LLC
Zarbeco MiScope Megapixel MP4K
with white LEDs
Synonym(s):
portable digital microscope, Handheld digital microcope, USB digital microscope, digital camera
About This Item
PDF Icon
Shipped & Sold By Zarbeco, LLC
A Trusted Marketplace Seller
PDF Icon
Have a question?
manufacturer/tradename
Zarbeco, LLC
General description
MiScope® Megapixel MP4K (MISC-MP4K) is a premium plus grade, lab quality, portable digital microscope used worldwide.  With its' micron level resolution, small footprint, easy to use included software, portability, and remote interface, it is a useful tool to supplement or even replace conventional stereo microscopes. Use your MIScope on-the-go or on valuable lab bench space and bring it to your sample, place it on top of it, and still pictures and movies can easily be saved, annotated, measured, drawn-on and enhanced with the included Video ToolBox Premier PC software. Powered by the USB-3 port of your laptop, desktop, or mobile device* it can go anywhere you need high quality images. Assembled in New Jersey by Zarbeco, LLC established in 2001.
  • 13 Megapixel camera
  • Resolution to 1.1 microns
  • 11x-360x magnification
  • Connects to the USB 3.0 port of your desktop or laptop computer, or mobile device*
  • Includes our Video Toolbox Premier PC Software for capturing still and moving images, for all PRO features plus side by side comparison, calibrated overlays, low-light frame averaging, enhanced measurement, pixel-level adjustment, magnifier window, and edge detection (MAC software optional MiTools on Mac App store).
  • Comes with variable Ushims for intermediate magnification.
  • Can be about 6-8 inches away from your object to see 1.9 inch field of view.
  • Move the arm on the side of your MiScope Megapixel MP4K and it will focus and zoom at the same time.  
  • When connected to the USB 3.0 port of your computer, it has fast image speeds at up to 60 frames per second without lag; 30 frames per second at 4K resolution with barely perceptible lag.
  • Has white LEDs with on-off switch to use side or bottom lighting with optional accessories.
*Apple iPad and iPhone need optional adapter. Apple, Mac, iPhone, and iPad are trademarks of Apple Inc., registered in the U.S. and other countries.
Legal Information
MiScope is a registered trademark of Zarbeco, LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)