011RM-250ML
Monobind Inc.
Monobind Inc. Drug-Free Human Urine
Raw & Pooled - 250 ml
Synonym(s):
Urine normal, urine drug free
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
liquid
storage temp.
< 8 °C
manufacturer/tradename
Monobind Inc.
General description
Human Urine pooled and processed into an optically clear and stable matrix. For use in preparing controls, calibrators and standards in various professional RUO-applications such as assay kits, lab analyses, research studies, and clinical trials.
- Shelf Life: 5 years from DOM at 8°C or Colder.
- Packaging & Shipping: 250ml in Media Bottle. Ships Frozen with Cool Packs, Overnight Only.
- Characteristics: Drug negative.
- Certificates of Analysis (COA): Ensures product has met all performance specifications prior to release for sale and accompanies every lot.
- Origin & Testing*: US Collection Centers - Donor Testing negative for (HIV-1&2, HCV, HBsAg, HIV-Ag) by FDA Method. *May vary by lot, refer to the provided COA.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.