AATB-21130
AAT Bioquest
AAT Bioquest Cal-520®, AM
10x50 ug
All Photos(1)
About This Item
Recommended Products
fluorescence
λex 492 nm;λem 515 nm
manufacturer/tradename
AAT Bioquest
General description
Cal-520® AM provides a robust homogeneous fluorescence-based assay tool for detecting intracellular calcium mobilization. The characteristics of its long wavelength, high sensitivity, and >100 times fluorescence enhancement, make Cal-520® AM an ideal indicator for the measurement of cellular calcium. The high S/N ratio and better intracellular retention make the Cal-520® calcium assay a robust tool for evaluating GPCR and calcium channel targets as well as for screening their agonists and antagonists.
Legal Information
Cal-520® is a trademark of AAT Bioquest
Choose from one of the most recent versions:
Certificates of Analysis (COA)