MilliporeSigma
All Photos(1)
Key Documents
688959
Sigma-Aldrich
(R,R)-Bicyclo[2.2.1]heptane-2,5-dione
98%
Sign Into View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C7H8O2
CAS Number:
Molecular Weight:
124.14
Beilstein/REAXYS Number:
9477671
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.22
assay
≥97.5% (GC)
98%
form
powder
optical activity
[α]/D 4.5±1.0°, c = 1 in chloroform
functional group
ketone
SMILES string
O=C1C[C@H]2C[C@@H]1CC2=O
InChI
1S/C7H8O2/c8-6-2-4-1-5(6)3-7(4)9/h4-5H,1-3H2/t4-,5-/m1/s1
InChI key
RVFBNLMXCXFOET-RFZPGFLSSA-N
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
pictograms
Exclamation mark
signalword
Warning
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library
Articles
Chiral diene ligands enable asymmetric transformations, constructing enantioenriched compounds from achiral substrates efficiently.
Questions
Reviews
No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service