Skip to Content
MilliporeSigma
Sodium cacodylate trihydrate purum p.a., ≥98.0% (T)

Sodium cacodylate trihydrate

Sodium cacodylate trihydrate purum p.a., ≥98.0% (T)
Synonym(s):
Cacodylic acid sodium salt trihydrate, Dimethylarsinic acid sodium salt, Dimethylarsonic acid sodium salt
Linear Formula:
(CH3)2AsO2Na · 3H2O
CAS Number:
Molecular Weight:
214.03
EC Number:

Products

Shipping
Product Category
Brand
Feature
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Application
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing