Skip to Content
US
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Silicon carbide
Linear Formula:
SiC
CAS Number:
409-21-2
Molecular Weight:
40.10
Products
Shipping
Ships Today (2)
Brand
Sigma-Aldrich (4)
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
357391
Silicon carbide,
−400 mesh particle size, ≥97.5%
Expand
View Pricing
378097
Silicon carbide,
-200 mesh particle size
Expand
View Pricing
594911
Silicon carbide,
nanopowder, <100 nm particle size
Expand
View Pricing
776742
Silicon carbide,
nanofiber, D <2.5 μm, L/D ≥ 20, 98% trace metals basis
Expand
View Pricing
933740
Silicon carbide,
nanopowder, ≥99% trace metals basis, 18 nm avg. part. size
Expand
View Pricing