Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V001581

Sigma-Aldrich

p-Toluenesulfonyl chloride

99%

Synonym(s):

Tosyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO2Cl
CAS Number:
Molecular Weight:
190.65
Beilstein/REAXYS Number:
607898
EC Number:
MDL number:
Pricing and availability is not currently available.

vapor pressure

1 mmHg ( 88 °C)

Quality Level

assay

99%

bp

134 °C/10 mmHg (lit.)

mp

65-69 °C (lit.)

SMILES string

Cc1ccc(cc1)S(Cl)(=O)=O

InChI

1S/C7H7ClO2S/c1-6-2-4-7(5-3-6)11(8,9)10/h2-5H,1H3

InChI key

YYROPELSRYBVMQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

p-Toluenesulfonyl chloride (tosyl chloride or p-TsCl) is an organic sulfonyl chloride mainly used to convert hydroxyl and amine groups into good leaving groups by forming sulfonates.[1]

Application

p-Toluenesulfonyl chloride can be used as:
  • An additive to enhance the yield of symmetrical biaryls via palladium chloride catalyzed homo-coupling of aryl boronic acids in the absence of ligands.[2]
  • A chlorine source for the α-chlorination of ketones in the presence of LDA.[3]
  • A reactant in the tosylation of alcohols and phenols in the presence of heteropoly acids.[1]
  • An activator for the reaction between 2-alkynylbenzaldoxime and phenols to form 1-aroxyisoquinolines in the presence of silver triflate.[4]
  • A catalyst for the solvent-free preparation of symmetrical bis(benzhydryl)ethers from benzhydrols.[5]

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

262.4 °F - closed cup

flash_point_c

128 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solvent-free and selective tosylation of alcohols and phenols with p-toluenesulfonyl chloride by heteropolyacids as highly efficient catalysts
Fazaeli R, et al.
Canadian Journal of Chemistry, 84(5), 812-818 (2006)
Facile synthesis of symmetrical bis (benzhydryl) ethers using p-toluenesulfonyl chloride under solvent-free conditions
Brahmachari G & Banerjee B
Organic and Medicinal Chemistry Letters, 3(1), 1-1 (2013)
Generation of 1-aroxyisoquinolines via a silver-catalyzed reaction of 2-alkynylbenzaldoxime with phenol in the presence of p-toluenesulfonyl chloride
Xiao Q, et al.
Tetrahedron, 69(25), 5119-5122 (2013)
Ligandless palladium chloride-catalyzed homo-coupling of arylboronic acids in aqueous media
Kabalka GW & Wang L.
Tetrahedron Letters, 43(16), 3067-3068 (2002)
α-Chlorination of ketones using p-toluenesulfonyl chloride.
Brummond KM and Gesenberg KD.
Tetrahedron Letters, 40(12), 2231-2234 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service