Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6122

Sigma-Aldrich

AK2 human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE)

Synonym(s):

ADK2, ATP-AMP phophotransferase, ATP-AMP transphosphorylase 2, Adenylate kinase 2, Adenylate monophosphate kinase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥95% (SDS-PAGE)

form

liquid

mol wt

28.6 kDa (259 aa, 1-239 aa + NT His Tag)

packaging

pkg of 100 μg

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... AK2(204)

General description

Adenylate kinase (AK; adenosine triphosphate-adenosine monophosphate [ATP-AMP] phosphotransferase) is a ubiquitous monomeric enzyme involved energy metabolism of prokaryotic and eukaryotic cells. Three isozymes (AK1, AK2 and AK3) are characterized in vertebrates. Expression of these isozymes is tissue-specific and developmentally regulated. AK2 is localized in the mitochondrial intermembrane space and may play a role in apoptosis.

Physical form

1 mg/mL solution in 20 mM Tris-HCl buffer (pH 7.5) containing 5 mM DTT and 20% glycerol.

Preparation Note

Centrifuge the vial prior to opening.

Other Notes

MGSSHHHHHH SSGLVPRGSH MAPSVPAAEP EYPKGIRAVL LGPPGAGKGT QAPRLAENFC VCHLATGDML RAMVASGSEL GKKLKATMDA GKLVSDEMVV ELIEKNLETP LCKNGFLLDG FPRTVRQAEM LDDLMEKRKE KLDSVIEFSI PDSLLIRRIT GRLIHPKSGR SYHEEFNPPK EPMKDDITGE PLIRRSDDNE KALKIRLQAY HTQTTPLIEY YRKRGIHSAI DASQTPDVVF ASILAAFSKA TCKDLVMFI

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ru-Ping Chen et al.
BMC molecular biology, 13, 31-31 (2012-10-02)
Adenylate kinase 2 (AK2) is a phosphotransferase that catalyzes the reversible reaction 2ADP(GDP) ↔ ATP(GTP) + AMP and influences cellular energy homeostasis. However, the role of AK2 in regulating cell proliferation remains unclear because AK2 has been reported to be involved in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service