Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

KEM0005

Sigma-Aldrich

Lambda Exonuclease

Ultra-pure enzyme for nucleic acid modifications

Sign Into View Organizational & Contract Pricing


About This Item

grade

Molecular Biology
for molecular biology

assay

>99% (SDS-PAGE)

form

buffered aqueous solution

specific activity

80,000 U/mg

concentration

5,000 U/mL

shipped in

dry ice

storage temp.

−20°C

General description

Lambda Exonuclease is a highly processive 5′ → 3′ double-stranded exonuclease that degrades one strand of the duplex. Lambda exonuclease can initiate at blunt DNA or DNA containing 3′ single-stranded overhangs. Lambda exonuclease has greatly reduced activity on non-phosphorylated DNA, single-stranded DNA and DNA having protruding 5′ single-stranded termini. Lambda exonuclease will no initiate at a nick or gap.

Application

Suitable for applications requiring a highly processive 5′ – 3′ exonuclease which is active on both blunt and 5′ recessed ends.

Features and Benefits

  • Ultra-purification process for ultimate enzyme performance
  • Highest quality specifications for ultimate product consistency
  • Undetectable DNA and nuclease contamination

Components

Supplied with:KEM0005B (10X Lambda Exo Reaction Buffer)

Unit Definition

1 unit is defined as the amount of enzyme required to produce 10 nmol of acid-soluble deoxyribonucleotide from double-stranded substrate in 30 minutes at 37° C.

Physical form

Supplied in 25 mM Tris-HCl, 50 mM NaCl, 1.0 mM DTT, 0.1 mM EDTA and 50% glycerol at pH7.5 @ 25° C.

Other Notes

Source of protein: Purified from a strain of E. coli that overexpresses the exonuclease gene from bacteriophage Lambda.
Unit size: 10,000 U

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laure Rittié et al.
Journal of cell communication and signaling, 2(1-2), 25-45 (2008-09-04)
Since molecular cloning has become routine laboratory technique, manufacturers offer countless sources of enzymes to generate and manipulate nucleic acids. Thus, selecting the appropriate enzyme for a specific task may seem difficult to the novice. This review aims at providing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service