Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A6652

Sigma-Aldrich

Atranorin

Synonym(s):

3-Formyl-2,4-dihydroxy-6-methylbenzoic acid 3-hydroxy-4-(methoxycarbonyl)-2,5-dimethylphenyl ester, Atranoric acid, Parmelin, Parmelin acid, Usnarin, Usnarin acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H18O8
CAS Number:
Molecular Weight:
374.34
EC Number:
MDL number:
UNSPSC Code:
12352103

description

Material is a mixture of atranorin and chloroatranorin

SMILES string

COC(=O)c1c(C)cc(OC(=O)c2c(C)cc(O)c(C=O)c2O)c(C)c1O

InChI

1S/C19H18O8/c1-8-5-12(21)11(7-20)17(23)15(8)19(25)27-13-6-9(2)14(18(24)26-4)16(22)10(13)3/h5-7,21-23H,1-4H3

InChI key

YLOYKYXNDHOHHT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Agnieszka Galanty et al.
Toxicology in vitro : an international journal published in association with BIBRA, 40, 161-169 (2017-01-18)
Lichens are an interesting source of potential anti-tumor compounds, among which usnic acid and atranorin seem to be the most promising, but their impact on invasive potential of tumor cells has not yet been comprehensively addressed. The aim of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service