Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

94820

Sigma-Aldrich

Variamine blue B salt

for microscopy (Hist.)

Synonym(s):

4-Amino-4′-methoxydiphenylamine-diazonium chloride, Diazo-4-amino-4′-methoxy-diphenylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H12ClN3O
CAS Number:
Molecular Weight:
261.71
Colour Index Number:
37255
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

grade

for microscopy (Hist.)

form

powder

SMILES string

[Cl-].COc1ccc(Nc2ccc(cc2)[N+]#N)cc1

InChI

1S/C13H12N3O.ClH/c1-17-13-8-6-11(7-9-13)15-10-2-4-12(16-14)5-3-10;/h2-9,15H,1H3;1H/q+1;/p-1

InChI key

AAXAHUUIRLOQMB-UHFFFAOYSA-M

Application

Variamine blue B salt is a chromogenic reagent/substrate used for assays that require the detection an oxidizing component.

pictograms

Skull and crossbonesCorrosion

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Basavaiah et al.
Farmaco (Societa chimica italiana : 1989), 58(4), 285-292 (2003-05-03)
Four simple, selective, accurate and reproducible procedures are described for the assay of methdilazine in bulk form and in formulations. One titrimetric and three spectrophotometric methods are based on the oxidation of the drug with potassium iodate, and determination of
H D Revanasiddappa et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 17(11), 1309-1312 (2002-01-05)
A rapid and sensitive spectrophotometric method is described for the determination of trace amounts of selenium using Variamine Blue (VB) as a chromogenic reagent. The proposed method is based on the reaction of selenium with potassium iodide in an acidic
Elmorsy Khaled et al.
Talanta, 83(5), 1538-1543 (2011-01-18)
Four sensitive catalytic potentiometric methods have been developed for trace levels determination of iodide based on its catalytic effects on the oxidation of four dyes: viz. variamine blue (VB), rhodamine B (RB), methylene blue (MB) and malachite green (MG), with
Demetrius G Themelis et al.
Talanta, 72(1), 277-281 (2007-04-15)
A highly selective and simple flow injection method is reported for the determination of Au(III) in jewel samples. The method is based on the catalytic effect of Au(III) on the oxidation of 4-amino-4'-methoxydiphenylamine hydrochloride (Variamine Blue B base, VB) by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service