65562
2-Picolylamine
derivatization grade (HPLC), LiChropur™, ≥98.0% (GC)
Synonym(s):
2-(Aminomethyl)pyridine
About This Item
Recommended Products
grade
derivatization grade (HPLC)
Quality Level
assay
≥98.0% (GC)
quality
LiChropur™
technique(s)
HPLC: suitable
refractive index
n20/D 1.544 (lit.)
n20/D 1.545-1.547
bp
82-85 °C/12 mmHg (lit.)
density
1.049 g/mL at 25 °C (lit.)
SMILES string
NCc1ccccn1
InChI
1S/C6H8N2/c7-5-6-3-1-2-4-8-6/h1-4H,5,7H2
InChI key
WOXFMYVTSLAQMO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Legal Information
signalword
Danger
hcodes
Hazard Classifications
Skin Corr. 1B
Storage Class
8A - Combustible corrosive hazardous materials
wgk_germany
WGK 3
flash_point_f
210.2 °F - closed cup
flash_point_c
99 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service