52793
(2S,5S)-2,5-Hexanediol
puriss., ≥99.0% (sum of enantiomers, GC)
About This Item
grade
puriss.
assay
≥99.0% (sum of enantiomers, GC)
optical activity
[α]20/D +35±2°, c = 9% in chloroform
optical purity
enantiomeric ratio: ≥99.5:0.5 (GC)
bp
212-215 °C (lit.)
mp
50-53 °C (lit.)
52-54 °C
storage temp.
2-8°C
SMILES string
C[C@H](O)CC[C@H](C)O
InChI
1S/C6H14O2/c1-5(7)3-4-6(2)8/h5-8H,3-4H2,1-2H3/t5-,6-/m0/s1
InChI key
OHMBHFSEKCCCBW-WDSKDSINSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
215.6 °F - closed cup
flash_point_c
102 °C - closed cup
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service