40870
1,2-Dimethyl-3-nitrobenzene
Selectophore™, ≥99.0%
Synonym(s):
3-Nitro-o-xylene
About This Item
Recommended Products
description
for ion-selective electrodes
Quality Level
product line
Selectophore™
assay
≥99.0% (GC)
≥99.0%
97%
form
liquid
refractive index
n20/D 1.543 (lit.)
bp
245 °C (lit.)
mp
7-9 °C (lit.)
density
1.129 g/mL at 25 °C (lit.)
SMILES string
Cc1cccc(c1C)[N+]([O-])=O
InChI
1S/C8H9NO2/c1-6-4-3-5-8(7(6)2)9(10)11/h3-5H,1-2H3
InChI key
FVHAWXWFPBPFOS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Legal Information
hcodes
pcodes
Hazard Classifications
Aquatic Chronic 2
Storage Class
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk_germany
WGK 2
flash_point_f
224.6 °F - closed cup
flash_point_c
107 °C - closed cup
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service