Recommended Products
grade
purum
assay
≥95.0% (GC)
refractive index
n20/D 1.468 (lit.)
n20/D 1.468
bp
149-150 °C/35 mmHg (lit.)
density
1.063 g/mL at 25 °C (lit.)
SMILES string
BrCCCCCCCCCC=C
InChI
1S/C11H21Br/c1-2-3-4-5-6-7-8-9-10-11-12/h2H,1,3-11H2
InChI key
YPLVPFUSXYSHJD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Replaced by
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
235.4 °F - closed cup
flash_point_c
113 °C - closed cup
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service