Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABN414

Sigma-Aldrich

Anti-LRRC4 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Leucine-rich repeat-containing protein 4, Brain tumor-associated protein BAG, Nasopharyngeal carcinoma-associated gene 14 protein, Netrin-G2 ligand, NGL-2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$377.00

$377.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$377.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$377.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat, human

species reactivity (predicted by homology)

canine (based on 100% sequence homology), feline (based on 100% sequence homology), bovine (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LRRC4(64101)

General description

Leucine-rich repeat-containing protein 4, or LRRC4, also known as Nasopharyngeal carcinoma-associated gene 14 protein or Netrin-G2 ligand (NGL-2), and encoded by the gene LRRC4/BAG, is a synaptic adhesion protein that regulates the formation of excitatory synapses through the recruitment of key pre and post synaptic proteins. LRRC4 is particularly important in auditory synaptic responses and LRRC4 is a key regulator of input-specific synapse development in the hippocampus as well. Finally, LRRC4 appears important in the suppression of glioma cancers in the brain and is specifically down regulated in tumors, and is currently being investigated as a potential anti-cancer target. LRRC4 is localized to the cell junctions and synapses. It is a cell membrane bound, post-synaptic, protein, and LRRC4’s expression is brain specific.

Immunogen

Epitope: Extracellular domain
KLH-conjugated linear peptide corresponding to the extracellular domain of human LRRC4.

Application

Anti-LRRC4 Antibody is a highly specific rabbit polyclonal antibody, that targets Leucine-rich repeat-containing protein 4 & has been tested in western blotting & IHC.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Lrrc4 in rat hippocampus tissue, whereas a 1:500 dilution from a representative lot detected LRRC4 in human cerebellum and thalamus tissues.
Research Category
Neuroscience
Research Sub Category
Synapse & Synaptic Biology

Quality

Evaluated by Western Blotting in human brain tissue lysate.

Western Blotting: 1 µg/mL of this antibody detected LRRC4 in 10 µg of human brain tissue lysate.

Target description

~100 kDa observed. This protein may be observed at higher molecular weights due to glycosylation.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service