Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

8.45002

Sigma-Aldrich

Maleic acid

Msynthplus

Synonym(s):

cis-Butenedioic acid, Toxilic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH=CHCO2H
CAS Number:
Molecular Weight:
116.07
Beilstein/REAXYS Number:
605762
MDL number:
UNSPSC Code:
12352106
EC Index Number:
203-742-5

vapor pressure

<0.1 hPa ( 20 °C)

assay

≥99% (acidimetric)

form

powder

potency

708 mg/kg LD50, oral (Rat)
1560 mg/kg LD50, skin (Rabbit)

pH

1.3 (20 °C, 100 g/L in H2O)

mp

130-135 °C (lit.)

solubility

478.8 g/L

density

1.59 g/mL at 25 °C (lit.)

bulk density

750‑800 kg/m3

storage temp.

2-30°C

SMILES string

OC(=O)\C=C/C(O)=O

InChI

1S/C4H4O4/c5-3(6)1-2-4(7)8/h1-2H,(H,5,6)(H,7,8)/b2-1-

InChI key

VZCYOOQTPOCHFL-UPHRSURJSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Colour (visual): white
Appearance of substance (visual): powder
Assay (HPLC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 130 °C
Melting range (upper value): ≤ 135 °C
Assay (acidimetric): ≥ 99.0 %
Identity (IR): passes test

Legal Information

MSYNTH is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service