Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.02590

Sigma-Aldrich

4-Chlorobutyryl chloride

for synthesis

Synonym(s):

4-Chlorobutyryl chloride, 4-Chlorobutyric acid chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl(CH2)3COCl
CAS Number:
Molecular Weight:
141.00
MDL number:
UNSPSC Code:
12352106
EC Index Number:
225-059-1

vapor pressure

1.31 hPa ( 20 °C)

Quality Level

assay

≥98.0% (GC)

autoignition temp.

440 °C

potency

1350 mg/kg LD50, oral (Rat)

expl. lim.

5.5-11.7 % (v/v)

bp

173-174 °C/1013 hPa

mp

-49 °C

transition temp

flash point 85 °C

density

1.258 g/cm3 at 20 °C

InChI

1S/C4H6Cl2O/c5-3-1-2-4(6)7/h1-3H2

InChI key

CDIIZULDSLKBKV-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.255 - 1.257
Identity (IR): passes test

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 1

flash_point_f

185.0 °F

flash_point_c

85 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service