Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W360805

Sigma-Aldrich

1-Hexen-3-ol

≥98%, stabilized, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH(OH)CH=CH2
CAS Number:
Molecular Weight:
100.16
FEMA Number:
3608
EC Number:
Council of Europe no.:
10220
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.104
NACRES:
NA.21

biological source

synthetic

grade

FG
Halal
Kosher

agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002

assay

≥98%

may contain

<0.10% alpha-tocopherol, synthetic as stabilizer

refractive index

n20/D 1.428 (lit.)

bp

134-135 °C (lit.)

density

0.834 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

documentation

see Safety & Documentation for available documents

food allergen

no known allergens

organoleptic

green; ethereal; rum

SMILES string

CCCC(O)C=C

InChI

1S/C6H12O/c1-3-5-6(7)4-2/h4,6-7H,2-3,5H2,1H3

InChI key

BVOSSZSHBZQJOI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

95.0 °F - closed cup

flash_point_c

35 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pingxi Xu et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(46), 16592-16597 (2014-10-29)
Insect repellents are important prophylactic tools for travelers and populations living in endemic areas of malaria, dengue, encephalitis, and other vector-borne diseases. DEET (N,N-diethyl-3-methylbenzamide) is a 6-decade-old synthetic repellent, which is still considered the gold standard of mosquito repellents. Mosquitoes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service