Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W213608

Sigma-Aldrich

Benzyl acetoacetate

≥97%

Synonym(s):

Acetoacetic acid benzyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2COOCH2C6H5
CAS Number:
Molecular Weight:
192.21
FEMA Number:
2136
Beilstein/REAXYS Number:
2049615
EC Number:
Council of Europe no.:
244
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.406

assay

≥97%

availability

not available in North America

refractive index

n20/D 1.512 (lit.)

bp

156-159 °C/10 mmHg (lit.)

density

1.112 g/mL at 25 °C (lit.)

SMILES string

CC(=O)CC(=O)OCc1ccccc1

InChI

1S/C11H12O3/c1-9(12)7-11(13)14-8-10-5-3-2-4-6-10/h2-6H,7-8H2,1H3

InChI key

WOFAGNLBCJWEOE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

comparable product

Product No.
Description
Pricing

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service