Skip to Content
MilliporeSigma
All Photos(1)

Documents

10898

Sigma-Aldrich

Benzyl acetoacetate

produced by Wacker Chemie AG, Burghausen, Germany, ≥96% (GC)

Synonym(s):

AC-benzyl, Acetoacetic acid benzyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2COOCH2C6H5
CAS Number:
Molecular Weight:
192.21
Beilstein/REAXYS Number:
2049615
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

produced by Wacker Chemie AG, Burghausen, Germany

assay

≥96% (GC)

manufacturer/tradename

Wacker Chemie AG

refractive index

n20/D 1.512 (lit.)

bp

156-159 °C/10 mmHg (lit.)

density

1.112 g/mL at 25 °C (lit.)

SMILES string

CC(=O)CC(=O)OCc1ccccc1

InChI

1S/C11H12O3/c1-9(12)7-11(13)14-8-10-5-3-2-4-6-10/h2-6H,7-8H2,1H3

InChI key

WOFAGNLBCJWEOE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzyl acetoacetate undergoes asymmetric bioreduction to form benzyl (S)-(+)-3-hydroxybutyrate in the presence of Candida schatavii strain MY 1831.

Other Notes

prices for bulk quantities on request

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asymmetric bioreduction of benzyl acetoacetate to its corresponding alcohol, benzyl (S)-(+)-3-hydroxybutyrate by the yeast Candida schatavii strain 1831.
Chartrain M, et al.
Journal of Fermentation and Bioengineering, 82(5), 507-508 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service