Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

89714

Sigma-Aldrich

(+)-(1S)-Menthyl (R)-p-toluenesulfinate

≥98.0% (sum of enantiomers, HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H26O2S
CAS Number:
Molecular Weight:
294.45
Beilstein/REAXYS Number:
4907352
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

assay

≥98.0% (sum of enantiomers, HPLC)

form

crystals

optical activity

[α]20/D +197±7°, c = 2% in acetone

mp

101-104 °C

shipped in

wet ice

storage temp.

−20°C

SMILES string

C[C@@H]1CC[C@@H](C(C)C)[C@@H](C1)OS(=O)c2ccc(C)cc2

InChI

1S/C17H26O2S/c1-12(2)16-10-7-14(4)11-17(16)19-20(18)15-8-5-13(3)6-9-15/h5-6,8-9,12,14,16-17H,7,10-11H2,1-4H3/t14-,16+,17-,20?/m1/s1

InChI key

NQICGNSARVCSGJ-IUINDACWSA-N

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service