Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

862088

Sigma-Aldrich

Lysergol

97%

Synonym(s):

9,10-Didehydro-6-methylergoline-8β-methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18N2O
CAS Number:
Molecular Weight:
254.33
EC Number:
MDL number:
UNSPSC Code:
12352200

assay

97%

SMILES string

CN1C[C@H](CO)C=C2[C@H]1Cc3c[nH]c4cccc2c34

Looking for similar products? Visit Product Comparison Guide

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 2 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anupam Maurya et al.
Biomedical chromatography : BMC, 26(9), 1096-1100 (2011-11-29)
A rapid, simple, sensitive, gradient and reproducible, reverse-phase high-performance liquid chromatographic method was developed for the quantitative estimation of bioactive alkaloids, lysergol and chanoclavine in the seeds of Ipomoea muricata. The clavine alkaloid, lysergol, is a bioenhancer for the drugs
D S Sveshnikov et al.
Aviakosmicheskaia i ekologicheskaia meditsina = Aerospace and environmental medicine, 33(5), 40-45 (1999-12-11)
Acute experiments with dogs showed that irritation of the sympathetic trunk in the thoracic cavity is more likely to amplify than inhibit duodenal contractions. This stimulating effect is better seen with alpha- and beta-adrenoreceptors blocked by phentolamine hydrochloride and inderal
V Kren et al.
Bioorganic & medicinal chemistry, 4(6), 869-876 (1996-06-01)
New glycosides derived from ergot alkaloids elymoclavine and DH-lysergol were synthesized by chemoenzymatic methods. beta-Glucosides were obtained either by chemical method or by transglycosylation (glycosidase from Aspergillus oryzae), lactosides were prepared by further extension of carbohydrate chain using beta-1,4-galactosyltransferase (bovine
S L Lodge-Ivey et al.
Journal of veterinary diagnostic investigation : official publication of the American Association of Veterinary Laboratory Diagnosticians, Inc, 18(4), 369-374 (2006-08-23)
Ergot alkaloids present in endophyte-infected tall fescue induce fescue toxicosis in livestock consuming the plant. The lysergic acid (LA) ring structure is a common moiety among the ergot alkaloids. Little is known about the bioavailability of LA because of limitations
Anupam Maurya et al.
Chemical biology & drug design, 81(4), 484-490 (2013-01-08)
Antibacterial activity of lysergol (1) and its semi-synthetic derivatives (2-14) and their synergy with the conventional antibiotic nalidixic acid (NA) against nalidixic acid-sensitive (NASEC) and nalidixic acid-resistant (NAREC) strains of Escherichia coli were evaluated. Lysergol (1) and derivatives (2-14) did

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service