Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

652660

Sigma-Aldrich

4-Bromo-3-methoxyaniline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3(OCH3)NH2
CAS Number:
Molecular Weight:
202.05
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
solid
assay:
97%

assay

97%

form

solid

mp

96-100 °C (lit.)

SMILES string

COc1cc(N)ccc1Br

InChI

1S/C7H8BrNO/c1-10-7-4-5(9)2-3-6(7)8/h2-4H,9H2,1H3

InChI key

RUTNWXBHRAIQSP-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service