Skip to Content
MilliporeSigma
All Photos(2)

Documents

418501

Sigma-Aldrich

Hexamethylenetetramine tribromide

97%

Synonym(s):

Hexamethylenetetramine hydrobromide perbromide, Hexamethylenetetramine, compound with hydrogen tribromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12N4 · HBr · Br2
CAS Number:
Molecular Weight:
380.91
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

220 °C (dec.) (lit.)

SMILES string

Br.BrBr.C1N2CN3CN1CN(C2)C3

InChI

1S/C6H12N4.Br2.BrH/c1-7-2-9-4-8(1)5-10(3-7)6-9;1-2;/h1-6H2;;1H

InChI key

CERSDEAKMGSGGO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monohalogenation (bromination and iodination) of electron rich arenes by tetraalkylammonillm halides in presence of cone. H2S04.
Myint YY and Pasha MA.
Indian Journal of Chemistry, 43, 357-360 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service