Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

335304

Sigma-Aldrich

(S)-(+)-4-Isopropyl-3-propionyl-2-oxazolidinone

98%

Synonym(s):

(S)-3-Propionyl-4-isopropyl-2-oxazolidinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15NO3
CAS Number:
Molecular Weight:
185.22
Beilstein/REAXYS Number:
3542849
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

liquid

optical activity

[α]25/D +93°, c = 8.7 in methylene chloride

refractive index

n20/D 1.464 (lit.)

bp

102-106 °C/0.75 mmHg (lit.)

density

1.094 g/mL at 25 °C (lit.)

SMILES string

CCC(=O)N1[C@H](COC1=O)C(C)C

InChI

1S/C9H15NO3/c1-4-8(11)10-7(6(2)3)5-13-9(10)12/h6-7H,4-5H2,1-3H3/t7-/m1/s1

InChI key

HOWPHXVPNNPSAZ-SSDOTTSWSA-N

Application

(S)-(+)-4-Isopropyl-3-propionyl-2-oxazolidinone can be used as a building block to prepare:
  • (25S)-cholestenoic-26-acid, a potent ligand for the hormonal DAF-12 receptor in Caenorhabditis elegans[1]
  • doliculide[2]

Other Notes

Versatile chiral auxiliary for asymmetric synthesis. For a recent review, see Aldrichimica Acta .[3]

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Iseki et al.
Chemical & pharmaceutical bulletin, 43(11), 1897-1901 (1995-11-01)
A new fluorinated analog of vitamin D2, 24-epi-26,26,26,27,27,27-hexafluoro- 1 alpha,25-dihydroxyvitamin D2, was efficiently synthesized starting from (R)-4-isopropyl-3-propionyl-2- oxazolidinone with high stereochemical control. In all four physiological test systems, the fluorinate vitamin D2 analog was found to be slightly less active
Stereoselective synthesis of the hormonally active (25 S)-δ7-dafachronic acid,(25 S)-δ4-dafachronic acid,(25 S)-dafachronic acid, and (25 S)-cholestenoic acid.
Martin R, et al.
Organic & Biomolecular Chemistry, 6(23), 4293-4295 (2008)
Enantioselective total synthesis of doliculide, a potent cytotoxic cyclodepsipeptide of marine origin and structure-cytotoxicity relationships of synthetic doliculide congeners
Hiroyuki I, et al.
Tetrahedron, 50(45), 12853-12882 (1994)
Ager, D.J., et al.
Aldrichimica Acta, 30, 3-3 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service