281573
(S)-(−)-1-Amino-2-(methoxymethyl)pyrrolidine
95%
Synonym(s):
SAMP
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
assay
95%
form
liquid
optical activity
[α]20/D −79°, neat
optical purity
ee: 97% (GLC)
refractive index
n20/D 1.465 (lit.)
bp
42 °C/1.8 mmHg (lit.)
density
0.97 g/mL at 25 °C (lit.)
functional group
ether
SMILES string
COC[C@@H]1CCCN1N
InChI
1S/C6H14N2O/c1-9-5-6-3-2-4-8(6)7/h6H,2-5,7H2,1H3/t6-/m0/s1
InChI key
BWSIKGOGLDNQBZ-LURJTMIESA-N
Looking for similar products? Visit Product Comparison Guide
Application
(S)-(-)-1-Amino-2-(methoxymethyl)pyrrolidine may be used as a chiral auxiliary in the asymmetric synthesis of (S)-2-ethylhexanoic acid and (S)-2-n-propyl-4-pentenoic acid. It can undergo 1,4-addition to (E)-alkenyl-cyclohexyl sulfonates in the presence of zinc bromide to form β-hydrazino-cyclohexyl sulfonates.
Undergoes condensation reactions with carbonyl compounds to produce hydrazones which are useful synthons in highly enantioselective syntheses.
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
161.6 °F - closed cup
flash_point_c
72 °C - closed cup
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Journal of agricultural and food chemistry, 67(11), 3168-3178 (2019-02-26)
In this study we report a secretory protein that was purified from Serratia marcescens strain S3 isolated from soil from the tobacco rhizosphere. Subsequent mass spectrometry and annotation characterized the protein as secretory alkaline metalloprotease (SAMP). SAMP plays a crucial
Synlett, 897-897 (1992)
Synthesis, 725-725 (1993)
Cellular and molecular gastroenterology and hepatology, 8(3), 427-446 (2019-06-11)
Crohn's disease (CD) is a debilitating inflammatory disorder that affects more than 1.6 million people in North America alone. Members of the tumor necrosis factor superfamily are key regulators of intestinal inflammation; specifically, tumor necrosis factor-like weak inducer of apoptosis
Inflammatory bowel diseases, 23(3), 392-403 (2017-02-02)
A large body of evidence supports a central role of TLR5 and its natural ligand, flagellin, in Crohn's disease (CD), with the precise mechanism(s) still unresolved. We investigated the role of flagellin/TLR5 in SAMP1/YitFc (SAMP) mice, a spontaneous model of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service