Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

246743

Supelco

Polyethylenimine on silica gel

beads, 20-60 mesh

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
23151817
PubChem Substance ID:
NACRES:
SB.52

form

beads

technique(s)

LPLC: suitable

matrix

silica

matrix active group

primary amine

particle size

20-60 mesh

pore size

60 Å pore size

capacity

1.0 meq/mL

separation technique

anion exchange

SMILES string

O[SiH3].C1CN1

Looking for similar products? Visit Product Comparison Guide

General description

Polyethylenimine (PEI) is polymer with positive charge density. It has various uses like in water purification, paper industry, to remove nucleic acids from protein solutions and many more. This polymer was first used in DNA and oligonucleotides delivery. It may also be used as transfection reagent.[1] Silica gels are chemically modified with PEI to be used as sorbent.[2] PEI stabilizes high surface area solid to adsorb CO2.[3]

Application

Ion-exchange resin
Polyethylenimine may be used in separation medium for TLC, paper chromatography and LPLC. Polyethylenimine on silica gel may be used for thin layer chromatography.

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

M Ghoul et al.
Water research, 37(4), 729-734 (2003-01-18)
The decontamination of synthetic Pb(II), Zn(II), Cd(II), Ni(II) solutions was investigated, using silica gels chemically modified with poly(ethyleneimine) (PEI) as sorbents. Two families of sorbents, i.e. silica/PEI and crosslinked silica/PEI, were prepared and characterized. Then the removal of metal ions
Nicola Gargiulo et al.
Journal of colloid and interface science, 367(1), 348-354 (2011-11-16)
Samples of porous, foam-like TUD (Technische Universität Delft)-1 mesoporous silica were functionalized with polyethylenimine and were used as a substrate for CO(2) adsorption. Produced solids were characterized by means of electron microscopy, thermogravimetric analysis, and N(2) adsorption/desorption at 77K, in
Polymeric Gene Delivery: Principles and Applications
Science (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service