About This Item
Recommended Products
vapor density
1.93 (vs air)
vapor pressure
1939 mmHg ( 21.1 °C)
assay
≥99.0% (GC)
autoignition temp.
723 °F
expl. lim.
9.3 %
impurities
butadiene
isobutane
isobutylene
bp
−6.3 °C (lit.)
mp
−185 °C (lit.)
SMILES string
CCC=C
InChI
1S/C4H8/c1-3-4-2/h3H,1,4H2,2H3
InChI key
VXNZUUAINFGPBY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Packaging
Other Notes
control valve
signalword
Danger
hcodes
Hazard Classifications
Flam. Gas 1A - Press. Gas Liquefied gas
Storage Class
2A - Gases
wgk_germany
nwg
flash_point_f
-112.0 °F - closed cup
flash_point_c
-80 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service