Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

159271

Sigma-Aldrich

Cyclobutylamine hydrochloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C4H7NH2 · HCl
CAS Number:
Molecular Weight:
107.58
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

mp

180-183 °C (lit.)

SMILES string

Cl.NC1CCC1

InChI

1S/C4H9N.ClH/c5-4-2-1-3-4;/h4H,1-3,5H2;1H

InChI key

NFAZOGXQOWEWBM-UHFFFAOYSA-N

application

Cyclobutylamine hydrochloride was used in the preparation of cyclobutylarachidonylamide.[1]

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Jarrahian et al.
Journal of neurochemistry, 74(6), 2597-2606 (2000-05-23)
Two putative endocannabinoids, N-arachidonylethanolamine (AEA) and 2-arachidonylglycerol, are inactivated by removal from the extracellular environment by a process that has the features of protein-mediated facilitated diffusion. We have synthesized and studied 22 N-linked analogues of arachidonylamide for the purpose of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service