Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

131385

Sigma-Aldrich

1-Phenethyl-4-piperidone

98%

Synonym(s):

1-(2-Phenylethyl)-4-piperidone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H17NO
CAS Number:
Molecular Weight:
203.28
Beilstein/REAXYS Number:
130302
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

drug control

Decreto Lei 15/93: Tabela IA (Portugal)

mp

57-60 °C (lit.)

solubility

ethanol: soluble, clear to hazy, light yellow to yellow (50 mg/mL)

SMILES string

O=C1CCN(CC1)CCc2ccccc2

InChI

1S/C13H17NO/c15-13-7-10-14(11-8-13)9-6-12-4-2-1-3-5-12/h1-5H,6-11H2

InChI key

YDJXNYNKKXZBMP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Phenethyl-4-piperidone was used in the preparation of phenethyl derivative of the propanamide. It was also used to synthesize l-Phenethylazacycloheptan-4-one.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and reactions of some azacycloheptan-4-ols.
Casy AF and Birnbaum H.
Journal of the Chemical Society, 983, 5130-5134 (1964)
Nadine Jagerovic et al.
Bioorganic & medicinal chemistry, 10(3), 817-827 (2002-01-30)
The synthesis of new fentanyl analogues in which the benzene ring of the propioanilido group has been replaced by phenylpyrazole is described. Antinociceptive activity was evaluated using the writhing and hot plate tests in mice. Two compounds, and, showed interesting
Synthesis and reactions of some azacycloheptan-4-ols.
Journal of the Chemical Society, 983, 5130-5134 (1964)
Jeremy R Canfield et al.
Journal of forensic sciences, 65(5), 1432-1442 (2020-04-30)
Eosin Y is a potential new color test for use in detecting illicit drugs that has not been extensively studied. In the present study, a variety of drugs of abuse and fentanyl analogues were tested to determine which drugs will

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service