Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900857

Sigma-Aldrich

LB Agar

Vetec, reagent grade

Synonym(s):

LB Broth with agar (Lennox)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105500
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service