Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900658

Sigma-Aldrich

Neocuproine

Vetec, reagent grade, 98%

Synonym(s):

2,9-Dimethyl-1,10-phenanthroline, DMPHEN

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12N2
CAS Number:
Molecular Weight:
208.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

SMILES string

Cc1ccc2ccc3ccc(C)nc3c2n1

InChI

1S/C14H12N2/c1-9-3-5-11-7-8-12-6-4-10(2)16-14(12)13(11)15-9/h3-8H,1-2H3

InChI key

IYRGXJIJGHOCFS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Nazeem et al.
Mutagenesis, 24(5), 413-418 (2009-06-10)
Plumbagin, a naphthoquinone derived from the medicinal plant Plumbago zeylanica has been shown to exert anticancer and anti-proliferative activities in cells in culture as well as animal tumor models. In our previous paper, we have reported the cytotoxic action of
Highly diastereoselective C(sp3)-C(sp) cross-coupling reactions between 1,3- and 1,4-substituted cyclohexylzinc reagents and bromoalkynes through remote stereocontrol.
Tobias Thaler et al.
Angewandte Chemie (International ed. in English), 50(9), 2174-2177 (2011-02-24)
Saliha Esin Celik et al.
Analytica chimica acta, 674(1), 79-88 (2010-07-20)
A novel on-line HPLC-cupric reducing antioxidant capacity (CUPRAC) method was developed for the selective determination of polyphenols (flavonoids, simple phenolic and hydroxycinnamic acids) in complex plant matrices. The method combines chromatographic separation, constituent analysis, and post-column identification of antioxidants in
Yan Wang et al.
Journal of the American Society for Mass Spectrometry, 19(9), 1353-1360 (2008-07-19)
S-nitrosylation of proteins serves an important role in regulating diverse cellular processes including signal transduction, DNA repair, and neurotransmission. Identification of S-nitrosylation sites is crucial for understanding the significance of this post-translational modification (PTM) in modulating the function of a
Mustafa Ozyürek et al.
Analytica chimica acta, 616(2), 196-206 (2008-05-17)
Hydroxyl radicals (OH) generated in the human body may play an important role in tissue injury at sites of inflammation in oxidative stress-originated diseases. As a more convenient, efficient, and less costly alternative to HPLC/electrochemical detection techniques and to the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service