Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900324

Sigma-Aldrich

Methotrexate hydrate

Vetec, reagent grade, 98%

Synonym(s):

4-Amino-10-methylfolic acid hydrate, L-4-Amino-N10-methylpteroylglutamic acid hydrate, L-Amethopterin hydrate, Antifolan hydrate, MTX hydrate, Methylaminopterin hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22N8O5 · xH2O
CAS Number:
Molecular Weight:
454.44 (anhydrous basis)
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

storage temp.

−20°C

SMILES string

[H]O[H].CN(Cc1cnc2nc(N)nc(N)c2n1)c3ccc(cc3)C(=O)N[C@@H](CCC(O)=O)C(O)=O

InChI

1S/C20H22N8O5.H2O/c1-28(9-11-8-23-17-15(24-11)16(21)26-20(22)27-17)12-4-2-10(3-5-12)18(31)25-13(19(32)33)6-7-14(29)30;/h2-5,8,13H,6-7,9H2,1H3,(H,25,31)(H,29,30)(H,32,33)(H4,21,22,23,26,27);1H2/t13-;/m0./s1

InChI key

FPJYMUQSRFJSEW-ZOWNYOTGSA-N

Gene Information

human ... DHFR(1719)

Looking for similar products? Visit Product Comparison Guide

Application

Potent inhibitor of dihydrofolate reductase and agent for antitumor studies. Use to inhibit dihydrofolate reductase in DHFR-based protein expression systems.
Potent inhibitor of dihydrofolate reductase and agent for antitumor studies. Use to inhibit dihydrofolate reductase in DHFR-based protein expression systems. Also effective in treatment of pyrimethamine-resistant Plasmodium vivax malaria parasites.
Potent inhibitor of dihydrofolate reductase and agent for antitumor studies. Use to inhibit dihydrofolate reductase in DHFR-based protein expression systems. Also shows immunosuppressive effects in, e.g., rheumatoid arthritis.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Josef Singer et al.
Molecular cancer therapeutics, 13(7), 1777-1790 (2014-04-24)
Passive immunotherapy with monoclonal antibodies represents a cornerstone of human anticancer therapies, but has not been established in veterinary medicine yet. As the tumor-associated antigen EGFR (ErbB-1) is highly conserved between humans and dogs, and considering the effectiveness of the
Methotrexate: new uses for an old drug.
Philip J Hashkes et al.
The Journal of pediatrics, 164(2), 231-236 (2013-11-30)
Ian Joseph Cohen et al.
Pediatric blood & cancer, 61(1), 7-10 (2013-09-17)
To determine the optimal time of folinic acid rescue after methotrexate (MTX) treatment in patients with ALL, we selected and evaluated relevant studies that included doses, rescue delay, and side effects. Rescue at 42-48 hours resulted in considerable toxicity, except
Johanna P Cremers et al.
Current opinion in pulmonary medicine, 19(5), 545-561 (2013-07-25)
Although glucocorticosteroids are considered the first-line treatment in sarcoidosis, refractory cases require alternatives, such as methotrexate (MTX). The aim of this study was to develop, on behalf of the World Association of Sarcoidosis and Other Granulomatous Disorders (WASOG), multinational evidence-based
Daniel H Solomon et al.
The American journal of medicine, 126(8), 730-730 (2013-07-28)
Elevated tumor necrosis factor (TNF)-α likely contributes to the excess cardiovascular risk observed in rheumatoid arthritis. We compared the cardiovascular risk in rheumatoid arthritis patients starting a TNF-α blocking agent versus a nonbiologic disease-modifying antirheumatic drug (nbDMARD). Subjects with rheumatoid

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service