Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V800447

Sigma-Aldrich

Zinc

AR, dust, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
EC Number:
MDL number:
UNSPSC Code:
12141750
PubChem Substance ID:
assay:
≥99.5%
grade:
AR
form:
dust
Pricing and availability is not currently available.

grade

AR

vapor pressure

1 mmHg ( 487 °C)

product line

Vetec

assay

≥99.5%

form

dust

resistivity

5.8 μΩ-cm, 20°C

bp

907 °C (lit.)

mp

420 °C (lit.)

density

7.133 g/mL at 25 °C (lit.)

cation traces

Fe: ≤0.005%
heavy metals (as Pb): ≤0.005%

SMILES string

[Zn]

InChI

1S/Zn

InChI key

HCHKCACWOHOZIP-UHFFFAOYSA-N

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Casper Groth et al.
Development (Cambridge, England), 140(14), 3018-3027 (2013-06-21)
Developmental patterning requires the precise interplay of numerous intercellular signaling pathways to ensure that cells are properly specified during tissue formation and organogenesis. The spatiotemporal function of the Notch signaling pathway is strongly influenced by the biosynthesis and intracellular trafficking
Michael Alvares et al.
Pediatrics, 132(1), e229-e232 (2013-06-05)
As food allergies become increasingly prevalent and testing methods to identify "food allergy" increase in number, the importance of careful diagnosis has become even more critical. Misdiagnosis of food allergy and inappropriate use of unproven testing modalities may lead to
Bikha Ram Devrajani et al.
JPMA. The Journal of the Pakistan Medical Association, 63(3), 369-373 (2013-08-07)
To determine the serum zinc level in patients with pneumonia. The descriptive cross-sectional study was conducted at the Liaquat University Hospital, Hyderabad, Pakistan, from September 2008 to February 2009. All patients above 12 years of age and of either gender
Lena Martin et al.
Archives of animal nutrition, 67(3), 248-261 (2013-06-08)
This study aimed at investigating the effect of dietary zinc on performance, jejunal brush border membrane enzyme activities and mRNA levels of enzymes and two zinc transporters in piglets. A total of 126 piglets were weaned at 26 ±1 days
I A Popov et al.
Molekuliarnaia biologiia, 47(3), 498-504 (2013-07-31)
Alzheimer's disease, is a lethal neurodegenerative pathology, characterized by the formation of soluble neurotoxic oligomers of the human amyloid-beta peptide Abeta which get accumulated forming polymeric extracellular aggregates (so-called amyloid plaques). The isomerized at aspartate 7 isoform of the human

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service