Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800424

Sigma-Aldrich

Thionyl chloride

LR, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SOCl2
CAS Number:
Molecular Weight:
118.97
Beilstein/REAXYS Number:
1209273
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay:
≥99.5%
grade:
LR
form:
liquid
Pricing and availability is not currently available.

grade

LR

vapor pressure

97 mmHg ( 20 °C)

product line

Vetec

assay

≥99.5%

form

liquid

refractive index

n20/D 1.518 (lit.)

bp

79 °C (lit.)

mp

−105 °C (lit.)

density

1.631 g/mL at 25 °C (lit.)

SMILES string

ClS(Cl)=O

InChI

1S/Cl2OS/c1-4(2)3

InChI key

FYSNRJHAOHDILO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1A - STOT SE 3

target_organs

Respiratory system

supp_hazards

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Karunanithy et al.
Journal of pharmacobio-dynamics, 12(8), 468-475 (1989-08-01)
The relationships between contraceptive activity, hydrophobic character and alkaline hydrolysis rates were studied in three homologous series of norethisterone and levonorgestrel esters. Hydrophobic character was expressed by the high-performance liquid chromatographic retention term, VR(w) or retention volume at 100% aqueous
Xiangshu Xiao et al.
The Journal of organic chemistry, 70(16), 6496-6498 (2005-07-30)
The indeno[1,2-c]isoquinolines are an important class of topoisomerase I inhibitors with anticancer activity. The condensation of Schiff bases with homophthalic anhydrides provides a mixture of cis- and trans-4-carboxy-3,4-dihydro-3-phenyl-1(2H)isoquinolones. Although the cis products can be readily converted to indeno[1,2-c]isoquinolines with thionyl
Je Seob Park et al.
ACS applied materials & interfaces, 3(2), 360-368 (2011-01-07)
In this study, we present a facile means of fabricating graphene thin films via layer-by-layer (LbL) assembly of charged graphene nanosheets (GS) based on electrostatic interactions. To this end, graphite oxide (GO) obtained from graphite powder using Hummers method is
S Aksoy et al.
Journal of biotechnology, 60(1-2), 37-46 (1998-05-08)
Poly(methyl methacrylate-acrylic acid) microspheres were prepared and the acid groups were activated by using either carbodiimide (CDI) or thionyl chloride (SOCl2). alpha-Amylase was covalently bound on these activated microspheres. The properties of the immobilized enzyme were investigated and compared with
N L Benoiton et al.
International journal of peptide and protein research, 43(4), 321-324 (1994-04-01)
Fmoc-glutamic acid is converted by thionyl chloride into the dichloride, which spontaneously cyclizes to Fmoc-pyroglutamyl chloride. The latter is stable to water. Pure Fmoc-pyroglutamyl chloride is obtained by washing the reaction mixture with water, which destroys uncyclized dichloride by converting

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service