Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V800215

Sigma-Aldrich

Hydroxylamine hydrochloride

LR, ≥98%

Synonym(s):

Hydroxylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2OH · HCl
CAS Number:
Molecular Weight:
69.49
Beilstein/REAXYS Number:
3539763
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay:
≥98%
grade:
LR
form:
crystals
Pricing and availability is not currently available.

grade

LR

vapor pressure

0.001 hPa ( 50 °C)

product line

Vetec

assay

≥98%

form

crystals

pH

2.5-3.5 (20 °C, 50 g/L)

mp

155-157 °C (dec.) (lit.)

density

1.67 g/mL at 25 °C (lit.)

SMILES string

Cl.NO

InChI

1S/ClH.H3NO/c;1-2/h1H;2H,1H2

Inchi Key

WTDHULULXKLSOZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

MAO inhibitor; inhibits platelet aggregation.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral

target_organs

spleen

Storage Class

4.1A - Other explosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wanderley R Bastos et al.
Environmental research, 140, 191-197 (2015-04-13)
The Madeira River is the largest tributary of the Amazon River Basin and one of the most impacted by artisanal gold-mining activities, deforestation for agricultural projects, and recent hydroelectric reservoirs. Total Hg (and methylmercury-MeHg) concentrations was determined in 3182 fish
Anke Bill et al.
PloS one, 9(6), e97973-e97973 (2014-06-03)
The human prostacyclin receptor (hIP receptor) is a seven-transmembrane G protein-coupled receptor (GPCR) that plays a critical role in vascular smooth muscle relaxation and platelet aggregation. hIP receptor dysfunction has been implicated in numerous cardiovascular abnormalities, including myocardial infarction, hypertension
Ayman M Atta et al.
International journal of molecular sciences, 16(4), 6911-6931 (2015-03-31)
In the present study, a new magnetic powder based on magnetite can be used as a petroleum crude oil collector. Amidoximes based on rosin as a natural product can be prepared from a reaction between hydroxylamine and rosin/acrylonitrile adducts. The
Marte S Dragset et al.
Antimicrobial agents and chemotherapy, 59(4), 2256-2264 (2015-02-04)
Efficient iron acquisition is crucial for the pathogenesis of Mycobacterium tuberculosis. Mycobacterial iron uptake and metabolism are therefore attractive targets for antitubercular drug development. Resistance mutations against a novel pyrazolopyrimidinone compound (PZP) that is active against M. tuberculosis have been
Yuan Zhao et al.
Small (Weinheim an der Bergstrasse, Germany), 10(22), 4770-4777 (2014-08-20)
Chiral plasmonic assemblies with strong and tunable chiroptical activity are emerging materials yet challenging to fabricate. Moreover, shell-programmed chiroptical regulation is really rare. Here, the chiroptical activity of core-shell (CS) nanoparticles (NPs) heterodimers (HDs) with different types and thicknesses of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service