Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PS734-10MG

Aldicarb

Synonym(s):

O-(Methylcarbamoyl)-2-methyl-2-(methylthio)propionaldehyd-oxime

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14N2O2S
CAS Number:
Molecular Weight:
190.26
Beilstein/REAXYS Number:
2209304
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

packaging

ampule of 10 mg

manufacturer/tradename

Chem Service, Inc. PS734-10MG

SMILES string

CNC(ON=CC(C)(C)SC)=O

InChI

1S/C7H14N2O2S/c1-7(2,12-4)5-9-11-6(10)8-3/h5H,1-4H3,(H,8,10)

InChI key

QGLZXHRNAYXIBU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Richard C Raabe et al.
PloS one, 9(8), e105999-e105999 (2014-08-28)
Alcohol addiction is a widespread societal problem, for which there are few treatments. There are significant genetic and environmental influences on abuse liability, and understanding these factors will be important for the identification of susceptible individuals and the development of
J L Burgess et al.
Archives of internal medicine, 154(2), 221-224 (1994-01-24)
Aldicarb is the most potent of the commercially available carbamate pesticides and is an unusual source of acute human poisonings. We present the case of a 43-year-old man exposed to aldicarb who developed severe cholinergic symptoms and progressive weakness requiring
Toxicology of aldicarb.
R L Baron et al.
Reviews of environmental contamination and toxicology, 105, 1-70 (1988-01-01)
Aldicarb contamination of groundwater.
H A Moye et al.
Reviews of environmental contamination and toxicology, 105, 99-146 (1988-01-01)
J F Risher et al.
Environmental health perspectives, 72, 267-281 (1987-06-01)
Aldicarb, 2-methyl-2-(methylthio)propionaldehyde-O-methylcarbamoyloxime, is an oxime carbamate insecticide manufactured by the Union Carbide Corporation and sold under the trade name Temik. It is a soil-applied systemic pesticide used against certain insects, mites, and nematodes, and is applied below the soil surface

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service