PS51
Dimethylarsinic acid
analytical standard
Synonym(s):
Cacodylic acid, Dimethylarsinic acid, Dimethylarsonic acid, Hydroxydimethylarsine oxide
About This Item
Recommended Products
grade
analytical standard
packaging
ampule of 500 mg
manufacturer/tradename
Chem Service, Inc. PS51
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
agriculture
environmental
format
neat
SMILES string
C[As](C)(O)=O
InChI
1S/C2H7AsO2/c1-3(2,4)5/h1-2H3,(H,4,5)
InChI key
OGGXGZAMXPVRFZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B
Storage Class
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service